Please login to your online account to display your discounted pricing

Alfa Aesar™ Biphenyl-3-boronic acid, 98%


Quantity 5g

Manufacturer: alfa aesar™  L1755206

 View more versions of this product

Catalog No. AAL1755206

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 198.03
Beilstein 2836311
Synonym 3-Phenylbenzeneboronic acid
Quantity 5g
Odor Odorless
Chemical Name or Material Biphenyl-3-boronic acid
Molecular Formula C12H11BO2
Health Hazard 1 Warning
Assay 98%
MDL Number MFCD01318102
CAS 5122-95-2
Melting Point 206° to 211°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Specifications

Chemical Name or Material Biphenyl-3-boronic acid
CAS 5122-95-2
Molecular Formula C12H11BO2
Formula Weight 198.03
MDL Number MFCD01318102
Beilstein 2836311
Synonym 3-Phenylbenzeneboronic acid

Applications
It is a biological reagent used as a boronate-assisted fluorogenic chemosensor. It is evaluated for pharmacological activity as fatty acid amide hydrolase inhibitors. It is a reactant involved in coupling with potassium cyanate, quinones, or fluorous tagged N-hydroxyphthalimide and direct C-H arylation of electron-deficient heterocycles.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents