Please login to your online account to display your discounted pricing

Alfa Aesar™ Biphenyl-3-boronic acid, 98%

$52.28 - $148.86

Chemical Identifiers

Chemical Name or Material Biphenyl-3-boronic acid
CAS 5122-95-2
Molecular Formula C12H11BO2
Formula Weight 198.03
MDL Number MFCD01318102
Beilstein 2836311
Synonym 3-Phenylbenzeneboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1755203

  SDS
alfa aesar™
L1755203
1g Each for $52.28

AAL1755206

  SDS
alfa aesar™
L1755206
5g Each for $148.86
Description & Specifications

Specifications

Formula Weight 198.03
Molecular Formula C12H11BO2
Chemical Name or Material Biphenyl-3-boronic acid
CAS 5122-95-2
MDL Number MFCD01318102
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 206° to 211°C
Beilstein 2836311
Odor Odorless
Assay 98%
Health Hazard 1 Warning
Quantity 1g
Synonym 3-Phenylbenzeneboronic acid

Applications
It is a biological reagent used as a boronate-assisted fluorogenic chemosensor. It is evaluated for pharmacological activity as fatty acid amide hydrolase inhibitors. It is a reactant involved in coupling with potassium cyanate, quinones, or fluorous tagged N-hydroxyphthalimide and direct C-H arylation of electron-deficient heterocycles.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.