Please login to your online account to display your discounted pricing

Alfa Aesar™ Biotinyl-[glu1]-gastrin I, Human, (Phosphorylated)

For research use

Specifications

Formulation Solid
Recombinant No
Storage Requirements -30° to -10°C
Source Escherichia coli
Product Type Biotinyl-[glu1]-gastrin I, Human, (phosphorylated)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66905EXD

 
alfa aesar™
J66905EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66905MCR

 
alfa aesar™
J66905MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formulation Solid
Recombinant No
Storage Requirements -30° to -10°C
Source Escherichia coli
Product Type Biotinyl-[glu1]-gastrin I, Human, (phosphorylated)
Format Solid
Regulatory Status RUO