Please login to your online account to display your discounted pricing

Alfa Aesar™ Biotinyl-[glu1]-gastrin I, Human, (Phosphorylated)


For research use

Manufacturer: alfa aesar™  J66905EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66905EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Formulation Solid
Recombinant No
Quantity 0.5mg
Storage Requirements -30° to -10°C
Source Escherichia coli
Regulatory Status RUO
Format Solid
Protein Length Biotin-glu-gly-Pro-Trp-Leu-glu-glu-glu-glu-glu-Ala-Tyr(PO{3}H{2})-gly-Trp-Met-Asp-Phe-NH{2}
Product Type Biotinyl-[glu1]-gastrin I, Human, (phosphorylated)