Biotin-Ubiquitin DFS Item
Biotin-Ubiquitin
Manufacturer:  Life Technologies LSPV4379
 View more versions of this product
Catalog No. PV4379
Description
Description
Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination. Due to the method in labeling the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.
For use in combination with other LanthaScreen™ products including LanthaScreen™ Tb-Ubiquitin (PV4375), Fluorescein-Ubiquitin (PV4377), or a LanthaScreen™ antibody such as Tb-anti-biotin (PV4749).
Specifications
Specifications
1.19mg/mL
Frozen
9.2kDa
10μg
For research use only. Not for use in diagnostic procedures
Biotin-Ubiquitin
Pharma and biopharma, target and lead identification and validation
Frozen
Biotin-Ubiquitin
–20°C
1 Each
Biotin-Ubiquitin
Documents
For Research Use Only. Not for use in diagnostic procedures.