Please login to your online account to display your discounted pricing

Alfa Aesar™ Biotin-Substance P


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66653LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66653LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Sequence Biotin-Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2
Storage Requirements -30° to -10°C
Quantity 5mg
Format Solid
Purification Method HPLC
Regulatory Status RUO
Product Type Biotin-Substance P
Molecular Weight 1573.94