Learn More
(+)-Biotin N-succinimidyl ester, 98%, Thermo Scientific Chemicals
In coupling reactions to amino acids, peptides and proteins through primary amines
Supplier: Thermo Scientific Chemicals 04477101
Description
(+)-Biotin N-succinimidyl ester is used in coupling reactions to amino acids, peptides and proteins through primary amines. It is used as amino reactive biotin reagent and in the preparation of biotinylated surfaces or polypeptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(+)-Biotin N-succinimidyl ester | |
212°C to 215°C | |
C14H19N3O5S | |
0.25 g | |
+-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester | |
YMXHPSHLTSZXKH-RVBZMBCESA-N | |
(2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate | |
6710714 | |
98% |
35013-72-0 | |
White | |
MFCD00078531 | |
4720781 | |
Soluble in Dimethylformamide ?50mg/mL (Stable for at least one month in dry DMF). Soluble in Dimethyl sulfoxide(DMSO). | |
C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3 | |
341.382 | |
341.4 | |
Powder |
Chemical Identifiers
35013-72-0 | |
341.382 | |
YMXHPSHLTSZXKH-RVBZMBCESA-N | |
6710714 | |
C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3 |
C14H19N3O5S | |
MFCD00078531 | |
+-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester | |
(2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only