missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+)-Biotin N-succinimidyl ester, 98%

In coupling reactions to amino acids, peptides and proteins through primary amines

Manufacturer:  Alfa Aesar™ 4477103

 View more versions of this product

Catalog No. AA4477103



Specifications

Specifications

(+)-Biotin N-succinimidyl ester
212°C to 215°C
35013-72-0
MFCD00078531
+-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester
C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3
341.382
341.4
98%
White
1g
C14H19N3O5S
4720781
YMXHPSHLTSZXKH-RVBZMBCESA-N
(2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate
6710714
Powder

Chemical Identifiers

35013-72-0
341.382
YMXHPSHLTSZXKH-RVBZMBCESA-N
6710714
C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3
C14H19N3O5S
MFCD00078531
+-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester
(2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate
Safety and Handling

Safety and Handling

Recommended Storage : 2°C to 8°C

SDS Documents