missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Biotin Mouse, Alexa Fluor 488, Clone: BK-1/39, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $262.68

Specifications

Antigen Biotin
Clone BK-1/39
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124662 Life Technologies
LS53989580
25μg Each for $143.88
501129103 Life Technologies
LS53989582
100μg Each for $262.68
Specifications

Description

This BK-1/39 monoclonal antibody reacts with biotin, a B-complex vitamin. As a coenzyme, biotin is involved in the metabolism of fatty acids, amino acids, and gluconeogenesis. In the laboratory, biotin is used to tag molecules of interest for biochemical or cell biological studies. This monoclonal antibody can be used to detect biotinylated proteins and antibodies, particularly if signal amplification is desired.
Description & Specifications

Specifications

Biotin
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
BK-1/39
Flow Cytometry
Alexa Fluor 488
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL