Biotin-Anti-His-Tag Antibody
Biotin-Anti-His-Tag Antibody
Manufacturer: Life Technologies LSPV6089
Description
Biotin-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using biotin negatively impacts protein activity. These antibodies can be used with our Kinase Tracers and Eu-Streptavidin to conduct LanthaScreen™ Eu Kinase Binding Assays. Learn More and See Which Kinases Have Been Validated Using This AssaySpecifications
Biochemical Kinase Assays, Drug Discovery and Development, Kinase Biology, LanthaScreen™ Kinase Activity Assays | |
Biotin | |
Frozen | |
25μg | |
–20°C | |
Primary |
0.5mg/mL | |
Biotin-Anti-His-Tag Antibody, 25μg, 0.5mg/mL Concentration, HIT2 Clone, 150kDa Molecular Weight, Biotin Conjugate, Used For Biochemical Kinase Assays, Drug Discovery and Development, Kinase Biology, LanthaScreen™ kinase binding assays | |
150kDa | |
RUO, For research use only | |
Primary | |
1 Each |
For Research Use Only. Not for use in diagnostic procedures.