Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Biotin Ab-2, Mouse Monoclonal Antibody

Detect both the free and protein-conjugated forms of biotin with Thermo Scientific™ Biotin Ab-2, Mouse Monoclonal Antibody.

$250.10 - $716.61

Specifications

Antigen Biotin Ab-2
Clone BTN.4
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1048P

 
thermo scientific™ lab vision™
MS1048P
200μg/mL 1mL Each for $627.51

MS1048P0

 
thermo scientific™ lab vision™
MS1048P0
200μg/mL 100µL Each for $250.10

MS1048P1

 
thermo scientific™ lab vision™
MS1048P1
200μg/mL 500µL Each for $427.39

MS1048P1ABX

 
thermo scientific™ lab vision™
MS1048P1ABX
1mg/mL 100µL Each for $551.94

MS1048PABX

 
thermo scientific™ lab vision™
MS1048PABX
1mg/mL 200µL Each for $716.61
Description & Specifications

Specifications

Antigen Biotin Ab-2
Clone BTN.4
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity All
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Epitope Tags
Type Primary
Immunogen Biotin conjugated to BSA
Isotype IgG2a kappa

Detect both the free and protein-conjugated forms of biotin with Thermo Scientific™ Biotin Ab-2, Mouse Monoclonal Antibody.

Ab-1 has a very high affinity for biotin and is excellent for use in various amplification techniques. It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.

Host Species: Mouse

Clone: BTN.4

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: Biotin conjugated to BSA

Recommended for:

  • Western Blotting
  • Confocal Microscopy
  • Flow Cytometry
  • Image Analysis
  • In situ Nucleic Acid Hybridization

Note:

USA: RUO; Int'l: RUO