Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Biotin Ab-2, Mouse Monoclonal Antibody


Detect both the free and protein-conjugated forms of biotin with Thermo Scientific™ Biotin Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1048P1ABX

 View more versions of this product

Catalog No. MS1048P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Regulatory Status RUO
Cross Reactivity All
Applications Western Blotting
Immunogen Biotin conjugated to BSA
Isotype IgG2a kappa
Clone BTN.4
Antigen Biotin Ab-2
Format Concentrated
Quantity 100µL
Research Discipline Epitope Tags
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Detect both the free and protein-conjugated forms of biotin with Thermo Scientific™ Biotin Ab-2, Mouse Monoclonal Antibody.

Ab-1 has a very high affinity for biotin and is excellent for use in various amplification techniques. It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.

Host Species: Mouse

Clone: BTN.4

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: Biotin conjugated to BSA

Recommended for:

  • Western Blotting
  • Confocal Microscopy
  • Flow Cytometry
  • Image Analysis
  • In situ Nucleic Acid Hybridization

Note:

USA: RUO; Int'l: RUO