Please login to your online account to display your discounted pricing

Synergy™ NEO HTS Multi-Mode Microplate Readers


With multiple parallel detectors for ultra-fast measurements and a dedicated filter-based system for live cell assays

Manufacturer: biotek instruments inc  NEOALPHAB

 View more versions of this product

Catalog No. BTNEOALPHAB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Light Source Xenon flash
Depth (English) 20.7 in.
Width (English) 15.4 in.
Height (English) 16.1 in.
Weight (Metric) 35kg
Includes With Alpha laser, fluorescence intensity, fluorescence polarization, time resolved fluorescence, UV-Vis absorbance, luminescence
Product Type Multi-Mode Microplate Reader

  • Compact footprint makes this system ideal for automation or benchtop implementation
  • Four PMT detectors including a dual PMT read head
  • Fastest plate stacker with a plate load/unload time of 6 sec.
  • Powerful 100mW laser-based excitation source for Alpha assays
  • Patented Hybrid optical system that includes top and bottom filter-based optics for high speed and performance; top and bottom monochromator optics for ultimate wavelength flexibility
  • Advanced 4-Zone incubator with temperature control to 65°C to cover the broadest range of incubated assays
  • Multi-directional bar code reader supports standard 1-D bar codes and 2-D bar codes
  • Fast reading speeds (6 seconds for 96-well plates, 11 seconds for 384-well plates)

  • Bar code-labeled filter cubes for fluorescence and Alpha detection for error-free parameter selection
  • Optional integrated plate stacker allows walk-away automation of short- or long-term assays
  • Powerful, easy-to-use, Gen5 2.0 Data Analysis Software

Model: NEOB
Synergy NEO HTS Multi-Mode Reader with:

  • Top dual PMT and bottom filter optics
  • Top and bottom monochromator optics
  • Fluorescence intensity
  • Fluorescence polarization
  • Time resolved fluorescence
  • UV-Vis absorbance and luminescence
Model: NEOALPHAB
Synergy NEO HTS Multi-Mode Reader with:
  • Top dual PMT and bottom filter optics
  • Top and bottom monochromator optics
  • Alpha laser
  • Fluorescence intensity
  • Fluorescence polarization
  • Time resolved fluorescence
  • UV-Vis absorbance and luminescence

For In Vitro diagnostic use.