Please login to your online account to display your discounted pricing

BioTek™ Synergy™ H4 Hybrid Microplate Reader

Combines two powerful detection systems, monochromator-based and filter-based, in one compact unit

Specifications

Accuracy ±1% at 2 O.D., typical
Dynamic Range 0 to 99,999 (fluorescence, luminescence); 0 to 4.0 O.D. (absorbance)
Electrical Requirements 100-240V 50/60Hz
Emission Wavelength Range 300 to 700nm (800nm with red extended option)
Excitation Wavelength Range 200 to 700nm
View More Specs
Products
Catalog Number Mfr. No. Detector Type Includes Price Quantity    

BTH4MLFAD

 
biotek™
H4MLFAD
Absorbance With injectors CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Accuracy ±1% at 2 O.D., typical
Dynamic Range 0 to 99,999 (fluorescence, luminescence); 0 to 4.0 O.D. (absorbance)
Electrical Requirements 100-240V 50/60Hz
Emission Wavelength Range 300 to 700nm (800nm with red extended option)
Excitation Wavelength Range 200 to 700nm
Hertz 50/60Hz
Interface Bidirectional RS-232
Light Source Tungsten halogen and xenon flash
Linearity >lt;1% from 0 to 3.0 O.D. (typical)
Sensitivity 2.5pg/mL SF; typical
Voltage 100-240V

Enjoy complete flexibilty and assay choice for current as well as future demands. Advanced multi-detection system is capable of performing an unlimited number of microplate-based assays.

  • Hybrid Technology™: Synergy H4 combines the sensitivity of a filter-based system with the convenience of monochromator-based optics to provide the broadest range of applications available on the market today.
  • Detection modes: Fluorescence Intensity, Time-Resolved Fluorescence, Fluorescence Polarization, AlphaScreen™/AlphaLISA, Luminescence, UV-Visible absorbance, FRET, TR-FRET, BRET, well area scanning and spectral scanning.
  • Modular and upgradeable architecture: Read modes are available as individual modules for cost-effectiveness and peace of mind.
  • Quadruple mono system and variable bandpass selection: Synergy H4 optics incorporates two double-grating monochromators. This quadruple design provides the best in spectral scanning performance and flexibility.
  • Deep blocking filters and dichroic mirrors: Synergy H4's filter/dichroic combination provides the best possible performance in fluorescence, time resolved fluorescence and fluorescence polarization applications.
  • Compatible with Take3 Plate with 2μL microspots: Enables low volume 260nm nucleic acid quantification

Synergy H4 Monochromator System

  • Uses two double-grating monochromators with variable bandpass selection
  • Highest stray light reflection, continuous wavelength selection, spectral scanning
  • Combines high performance with convenience and flexibility
Synergy H4 Filter/Mirror System
  • Delivers more energy to the sample and provides high signal-to-noise rations
  • Faster read speed, more sensitivity, more precise control over optical parameters

CE Marked, ETL Listed