Please login to your online account to display your discounted pricing

BioTek™ Synergy™ H1 Hybrid Multi-Mode Monochromator Fluorescence Microplate Readers


Flexible monochromator-based detection with high performance dichroic-based filter detection. BioTek Synergy H1 Hybrid Multi-Mode Monochromator Fluorescence Microplate Readers with its monochromator optics supports top and bottom fluorescence intensity, UV-visible absorbance and high-performance luminescence detection.

Manufacturer: biotek™  BTH1MD

 View more versions of this product

Catalog No. 11-120-534

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multi-Mode Microplate Reader with Dispenser
Depth (Metric) 47.2cm
Weight (Metric) 22.5kg
Width (Metric) 39.1cm
Width (English) 15.4 in.
Depth (English) 18.6 in.
Height (English) 12.9 in.
Height (Metric) 32.8cm
Includes Monochromator fluorescence; Monochromator absorbance; Full-light luminescence; Dual reagent dispenser
Weight (English) 50 lb.
Light Source Xenon flash
No. of Dispensers 2.0

  • Quadruple grating monochromator for maximum flexibility and ease of use
  • Compatible with Take3™ Multi-Volume Plate with 16 microspots of 2μL each for low-volume nucleic acid quantification
  • Dichroic-based filter optics for best performance and advanced detection technologies, such as fluorescence polarization and time-resolved fluorescence
  • Third generation quadruple grating design of the monochromator optics allows working at any excitation or emission wavelength with a 1nm step
  • Filter module includes its own light source and a high-performance dichroic-based wavelength selection system
  • Dual reagent injection system option automates ion channel assays and flash luminescent assays
  • Available with Gen5™ Data Analysis Software: reader control, advanced data analysis and flexible Excel export in one software package

Accessories

  • Take3 Multi-Volume Plate
  • BioStack™: 30 or 50 plate stacker
  • Gen5 Secure for 21CFR part 11 compliance
  • Fluorescence, Luminescence and Absorbance Test Plates

H1MFDG

  • Compatible with gas controller

Note

  • Dual reagent dispenser option available with all models
Applications
  • Academic Life Science Research
  • Biotech industry—especially smaller companies and start-ups
  • Clinical—Filter module for emerging applications of TRF, FP and luminescence