Please login to your online account to display your discounted pricing

BioTek™ Synergy™ H1 Hybrid Multi-Mode Filter/Dichroic Fluorescence Microplate Readers

Flexible monochromator-based detection with high performance dichroic-based filter detection. BioTek Synergy H1 Hybrid Multi-Mode Filter/Dichroic Fluorescence Microplate Readers with its monochromator optics supports top and bottom fluorescence intensity, UV-visible absorbance and high-performance luminescence detection.

Specifications

Light Source Xenon flash
View More Specs
Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    

11-120-531

 
biotek™
BTH1F
Filter/dichroic fluorescence; Fluorescence polarization; Time-resolved fluorescence; Filtered luminescence Filter/Dichroic Fluorescence Microplate Reader CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

11-120-532

 
biotek instruments inc
BTH1FD
Filter/dichroic fluorescence; Fluorescence polarization; Time resolved fluorescence; Filtered luminescence; Dual reagent dispenser Filter/Dichroic Fluorescence Microplate Reader with Dispenser CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Light Source Xenon flash

  • Quadruple grating monochromator for maximum flexibility and ease of use
  • Compatible with Take3™ Multi-Volume Plate with 16 microspots of 2μL each for low-volume nucleic acid quantification
  • Dichroic-based filter optics for best performance and advanced detection technologies, such as fluorescence polarization and time-resolved fluorescence
  • Third generation quadruple grating design of monochromator optics allows working at any excitation or emission wavelength with 1nm step
  • Filter module includes its own light source and high-performance dichroic-based wavelength selection system
  • Dual reagent injection system option automates ion channel assays and flash luminescent assays
  • Available with Gen5™ Data Analysis Software: reader control, advanced data analysis and flexible Excel export in one software package

Accessories

  • Take3 Multi-Volume Plate
  • BioStack™: 30 or 50 plate stacker
  • Gen5 Secure for 21CFR part 11 compliance
  • Fluorescence, Luminescence and Absorbance Test Plates

H1MFDG

  • Compatible with gas controller

Note

  • Dual reagent dispenser option available with all models
Applications
  • Academic Life Science Research
  • Biotech industry—especially smaller companies and start-ups
  • Clinical—Filter module for emerging applications of TRF, FP and luminescence