Please login to your online account to display your discounted pricing

Synergy™ H1 Hybrid Multi-Mode Filter/Dichroic Fluorescence Microplate Readers


Flexible monochromator-based detection with high performance dichroic-based filter detection. BioTek Synergy H1 Hybrid Multi-Mode Filter/Dichroic Fluorescence Microplate Readers with its monochromator optics supports top and bottom fluorescence intensity, UV-visible absorbance and high-performance luminescence detection.

Manufacturer: biotek instruments inc  BTH1FD

 View more versions of this product

Catalog No. 11-120-532

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Filter/Dichroic Fluorescence Microplate Reader with Dispenser
Weight (Metric) 22.5kg
Depth (Metric) 47.2cm
Depth (English) 18.6 in.
Width (English) 15.4 in.
No. of Dispensers 2.0
Height (English) 12.9 in.
Width (Metric) 39.1cm
Weight (English) 50 lb.
Light Source Xenon flash
Height (Metric) 32.8cm
Includes Filter/dichroic fluorescence; Fluorescence polarization; Time resolved fluorescence; Filtered luminescence; Dual reagent dispenser