missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BioTek™ Synergy™ H1 Hybrid Multi-Mode All Detection Modes Microplate Readers

Flexible monochromator-based detection with high performance dichroic-based filter detection. BioTek Synergy H1 Hybrid Multi-Mode All Detection Modes Microplate Readers have monochromator optics which supports top and bottom fluorescence intensity, UV-visible absorbance and high-performance luminescence detection.

Specifications

Light Source Xenon flash
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type Price Quantity  
Catalog Number Mfr. No. Includes Product Type Price Quantity  
11-120-535 BioTek™
BTH1MF
Monochromator fluorescence; Monochromator Abs; Full-light luminescence; Filter/dichroic fluorescence; Fluorescence polarization; Time resolved fluorescence; Filtered luminescence Microplate Reader N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTH1MFDG BioTek™
H1MFDG
Monochromator fluorescence, full-light luminescence, filter/dichroic, time resolved fluorescence, polarization, filtered luminescence; Dual reagent dispenser; Compatible w/Gas controller Microplate Reader; Dispenser; Compatible w/ Gas Controller N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Quadruple grating monochromator for maximum flexibility and ease of use
  • Compatible with Take3™ Multi-Volume Plate with 16 microspots of 2μL each for low-volume nucleic acid quantification
  • Dichroic-based filter optics for best performance and advanced detection technologies, such as fluorescence polarization and time-resolved fluorescence
  • Third generation quadruple grating design of the monochromator optics allows working at any excitation or emission wavelength with a 1nm step
  • Filter module includes its own light source and a high-performance dichroic-based wavelength selection system
  • Dual reagent injection system option automates ion channel assays and flash luminescent assays
  • Available with Gen5™ Data Analysis Software: reader control, advanced data analysis and flexible Excel export in one software package

Accessories

  • Take3 Multi-Volume Plate
  • BioStack™: 30 or 50 plate stacker
  • Gen5 Secure for 21CFR part 11 compliance
  • Fluorescence, Luminescence and Absorbance Test Plates

H1MFDG

  • Gas controller compatible
  • Gas module must be purchased separately (BT1210007 or BT12310008)

Note

  • Dual reagent dispenser option available with all models

Applications

  • Academic Life Science Research
  • Biotech industry—especially smaller companies and start-ups
  • Clinical—Filter module for emerging applications of TRF, FP and luminescence

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Xenon flash