Please login to your online account to display your discounted pricing

BioTek™ Synergy™ H1 Hybrid Multi-Mode All Detection Modes Microplate Readers


Flexible monochromator-based detection with high performance dichroic-based filter detection. BioTek Synergy H1 Hybrid Multi-Mode All Detection Modes Microplate Readers have monochromator optics which supports top and bottom fluorescence intensity, UV-visible absorbance and high-performance luminescence detection.

Manufacturer: biotek™  H1MFDG

 View more versions of this product

Catalog No. BTH1MFDG

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Microplate Reader; Dispenser; Compatible w/ Gas Controller
Height (Metric) 32.8cm
Height (English) 12.9 in.
Light Source Xenon flash
Depth (Metric) 47.2cm
Includes Monochromator fluorescence, full-light luminescence, filter/dichroic, time resolved fluorescence, polarization, filtered luminescence; Dual reagent dispenser; Compatible w/Gas controller

  • Quadruple grating monochromator for maximum flexibility and ease of use
  • Compatible with Take3™ Multi-Volume Plate with 16 microspots of 2μL each for low-volume nucleic acid quantification
  • Dichroic-based filter optics for best performance and advanced detection technologies, such as fluorescence polarization and time-resolved fluorescence
  • Third generation quadruple grating design of the monochromator optics allows working at any excitation or emission wavelength with a 1nm step
  • Filter module includes its own light source and a high-performance dichroic-based wavelength selection system
  • Dual reagent injection system option automates ion channel assays and flash luminescent assays
  • Available with Gen5™ Data Analysis Software: reader control, advanced data analysis and flexible Excel export in one software package

Accessories

  • Take3 Multi-Volume Plate
  • BioStack™: 30 or 50 plate stacker
  • Gen5 Secure for 21CFR part 11 compliance
  • Fluorescence, Luminescence and Absorbance Test Plates

H1MFDG

  • Gas controller compatible
  • Gas module must be purchased separately (BT1210007 or BT12310008)

Note

  • Dual reagent dispenser option available with all models

Applications

  • Academic Life Science Research
  • Biotech industry—especially smaller companies and start-ups
  • Clinical—Filter module for emerging applications of TRF, FP and luminescence