missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BioTek™ Synergy™ 2 Multi-Mode Microplate Readers

Designed for life science and drug discovery applications. BioTek Synergy 2 Multi-Mode Microplate Readers include dedicated optical elements which are optimized for each specific modality for low limits of detection in one compact instrument.

Specifications

Depth (English) 17.5 in.
Depth (Metric) 44.5cm
Measurement Speed 96 wells: 11 seconds; 384 wells: 22 seconds; 1536 wells: 42 seconds
Dynamic Range 0 to 4.0 O.D.
Height (English) 14.5 in.
View More Specs

Includes: Gen5™ software controlled through USB or serial port


Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
36-101-5199 BioTek™
SLFPTAD
With all detection modes and injector N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
36-101-5200 BioTek™
SLFPA
With fluorescence intensity and polarization, luminescence, UV-visible absorbance N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT-SLFD BioTek™
SLFD
With fluorescence and luminescence N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
11-120-516 BioTek™
SLFA
With luminescence, fluorescence, absorbance N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
11-120-517 BioTek™
SLFAD
With luminescence, fluorescence, absorbance and injector N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
36-101-5201 BioTek™
SLFPTA
With all detection modes N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Fluorescence intensity, time resolved fluorescence, fluorescence polarization, glow and flash luminescence, UV-Vis absorbance, FRET, TR-FRET, BRET, area scanning, adn spectral scanning
  • Optical system includes a narrow band monochromator, optimized filter/dichroic mirror pairing and three broad spectrum light sources — SQ Xenon Flash, Tungsten Halogen and High Energy DPR Xenon Flash
  • Modular design allows for easily upgrading elements
  • Optimized throughput for screening assays in 6- to 1536-well plates (luminescence 6-384) and PCR plates
  • Compatible with Take3 and Take3 Trio Micro-Volume plates — up to 16 or 48 2μL samples for low-volume DNA, RNA and protein quantification
  • Temperature control from 4° above ambient to 50°C
  • 11-120-515 includes two syringe pumps with dispensing volume of 5-1000μL in 1μL increments

Applications

  • For in vitro diagnostic use (except in the European Union)
  • Screening assays; Binding assays; Ion channel assays; Quantitative assays (DNA, protein); Kinetic assays; Gene expression assays (GFP, Luciferase); ELISA assays; Cell proliferation, Cytotoxicity

Compliance

CE and NRTL Safety Agency marked

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

17.5 in.
96 wells: 11 seconds; 384 wells: 22 seconds; 1536 wells: 42 seconds
14.5 in.
0.001 O.D.
44.5cm
0 to 4.0 O.D.
37.3cm