BioTek™ Synergy™ 2 Multi-Mode Microplate Readers
Designed for life science and drug discovery applications. BioTek Synergy 2 Multi-Mode Microplate Readers include dedicated optical elements which are optimized for each specific modality for low limits of detection in one compact instrument.
Manufacturer:  BioTek™ SLFD
 View more versions of this product
Catalog No. BT-SLFD
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes: Gen5™ software controlled through USB or serial port
Specifications
Description
Microplate reader, Multi-Detection; BioTek Synergy 2; With fluorescence and luminescence; With dispenser; With Gen5 Reader Control and Data Analysis Software
  • Fluorescence intensity, time resolved fluorescence, fluorescence polarization, glow and flash luminescence, UV-Vis absorbance, FRET, TR-FRET, BRET, area scanning, adn spectral scanning
  • Optical system includes a narrow band monochromator, optimized filter/dichroic mirror pairing and three broad spectrum light sources — SQ Xenon Flash, Tungsten Halogen and High Energy DPR Xenon Flash
  • Modular design allows for easily upgrading elements
  • Optimized throughput for screening assays in 6- to 1536-well plates (luminescence 6-384) and PCR plates
  • Compatible with Take3 and Take3 Trio Micro-Volume plates — up to 16 or 48 2μL samples for low-volume DNA, RNA and protein quantification
  • Temperature control from 4° above ambient to 50°C
  • 11-120-515 includes two syringe pumps with dispensing volume of 5-1000μL in 1μL increments
Applications
  • For in vitro diagnostic use (except in the European Union)
  • Screening assays; Binding assays; Ion channel assays; Quantitative assays (DNA, protein); Kinetic assays; Gene expression assays (GFP, Luciferase); ELISA assays; Cell proliferation, Cytotoxicity
Compliance
CE and NRTL Safety Agency marked
Description & Specifications
Specifications
With fluorescence and luminescence
Gen5 Reader Control and Data Analysis Software