Please login to your online account to display your discounted pricing

BioTek™ Precision™ Microplate Pipetting System

Innovative replacement for manual pipetting

Specifications

Amperage ≤0.7A
Certifications/Compliance CE, TUV
Depth (English) 15 in.
Depth (Metric) 38.1cm
Dispensing Accuracy 1% typical at 100μL
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

BTPRC384U

 
biotek™
PRC384U
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTPRC384M

 
biotek™
PRC384M
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Amperage ≤0.7A
Certifications/Compliance CE, TUV
Depth (English) 15 in.
Depth (Metric) 38.1cm
Dispensing Accuracy 1% typical at 100μL
Dispensing Precision ≤1.5% CV typical at 100μL
Dispensing Speed 100μL/well, 96-wells, 1 x 8 : 14 sec., 100μL/well, 96-wells, single-channel : 4 min.
Hertz 50/60Hz
Plate Type 96 and 384-well microplates
Voltage 100 to 240V
Weight (English) 28 lb.
Weight (Metric) 12.7kg
Electrical Requirements 100 to 240V 50/60Hz
Height (English) 16 in.
Height (Metric) 40.64cm
Humidity 80%
Includes Manifold
Interface External membrane keypad with 25 alphanumeric keys, two-line x 24-character LCD
Operating Temperature Range (English) 59° to 104°F
Operating Temperature Range (Metric) 15° to 40°C
Software Precision Power™, for PC protocol programming and execution
Volume (English) Bottle 67.62 oz.
Volume (Metric) Bottle 2L
Volume (Metric) Range 5 to 120μL
Width (English) 21 in.

Features

  • Available with multichannel manifold for continuous reagent dispensing
  • Ideal for routine liquid deliveries such as reagent addition, plate replication, serial dilutions, sample transfer, and mixing within a well Configurable multistation platform and XY transport design provide effortless 96- to 384-well plate transfers
  • Proprietary pipet technology provides reliable way to pick up and seal standard tips with controlled force Open design guarantees compatibility with robotic plate handlers
  • Accommodates BioTek and other standard 50 to 250μL pipet tips; and 96- and 384-well flat-, round-, and V-bottom plates

Software

  • Preinstalled software enables user to copy existing demonstration protocol and modify it; memory stores up to 80 program files User can create, edit, copy, and delete files using external keypad and 24-character LCD — virtually any liquid transfer parameter can be customized Configurable aspiration depth ensures that tips don't touch bottom of wells while performing sensitive cell assays
  • Optional Precision Power™ graphical user interface (PRCPWRV2) simplifies program creation under Windows™ 98, NT, 2000, and XP
  • Programs can be created and run from a PC, or downloaded and run from onboard microprocessor

CAN/CSA approved

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.