BioTek™ Microplate Washer Flat Magnets
For insertion into BioTek washer biomagnetic separation carriers
Manufacturer:  BioTek™ 7103016
 View more versions of this product
Catalog No. BT7103016
Specifications Description & Specifications
Specifications
96-Well Flat Magnet
405 ls, 405 touch, el406, elx405, elx50 microplate washers