Please login to your online account to display your discounted pricing

BioTek™ Accessories for BioTek Microplate Readers


For use with Multi-mode, absorbance and monochromator based microplate readers.

Manufacturer: biotek™  75034

 View more versions of this product

Catalog No. BT75034

  • / Each
 

Description & Specifications

Specifications

Product Type Serial Cable; Modem (9-pin female to 9-pin male)

Serial Cable (BT75034)

  • Modem (9-pin female to 9-pin male)
  • For use with ELx800, Synergy™ 2, Synergy 2 Alpha, Synergy 2 SL, Synergy H1m, Synergy HT, Synergy Mx, Synergy MxF
Serial Cable (BT75053)
  • Null Modem (9-pin female to 25-pin female)
  • For use with ELx808™, FLx800, PowerWave, Precision, Precision XS, Synergy HT
Fluorescence Filter (BT7082225)
  • 590/20nm
  • For use with Cytation3, FLx800, Synergy 2, Synergy H1, Synergy H4, Synergy HT
Fuse (BT46024)
  • 250V 1.5A
  • For use with ELx808
Fuse (BT46023)
  • 0.75A
  • For use with PowerWave X