missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes

Dispense Cassettes for use with MultiFlo FX Dispenser and EL406 Washer Dispenser.

Supplier:  Agilent BioTek 7170010

 View more versions of this product

Catalog No. 11-120-627



Description

Description

  • Volume range 1 μL to 10 mL
  • Accommodates 6- to 1536-well microplates, PCR trays, deep-well microplates, microtubes and other tube configurations up to 4 in. in height
  • Dispense 1536 wells in 18 s.
  • Cassettes can be autoclaved without recalibration
  • Dispense up to eight reagents at one time
  • Customizable dispense pattern allows skipped columns or rows
  • Built-in priming trough
  • Easy maintenance of individually replaceable tips and choice of replacement cassettes or tubing
  • Compatible with Bio-Stack microplate stacker 11-120-601 for increased throughput

Specifications

Specifications

0.014 in.
For liquid handling
0.035 cm
Molded Polypropylene
10 to 3000 μL
Large Volume Cassette
10 μL
EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser
SDS
Documents

Documents

Promotions

Promotions