BioTek™ MicroFlo™ Select Dispenser
Choose dynamic dispense volume, microplate types, tube sizes and dispense height
Manufacturer:  BioTek™ 7170008
 View more versions of this product
Catalog No. 11-120-630
Specifications Description & Specifications
Specifications
Medium Volume Replacement Tubing Set
0.035cm
For liquid handling
For el406, multiflo™, multiflo™ fx
5 to 2500μL
0.014 in.
Molded Polypropylene
5μL