Please login to your online account to display your discounted pricing

BioTek™ FLx800™ Microplate Fluorescence Readers


Choose from models with top probe or top and bottom probes.

Manufacturer: biotek™  FLX800TBI

 View more versions of this product

Probe(s), 485 and 360nm excitation filters, 528 and 460nm emission filters, serial and power cables, power cord

Catalog No. BTFLX800TBI

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Emission Wavelength Range 340 to 650nm
Software Type Gen5 data analysis software
Excitation Wavelength Range 300 to 650nm
Excitation Filters 485 and 360 excitation filters
Emission Filters 528 and 460 emission filters
Dynamic Range 5 decades (0 to 99,999)
Incubator Temperature To 50°C
Width (Metric) 39cm
Width (English) 15.5 in.
Height (Metric) 23cm
Height (English) 9 in.
Hertz 50/60Hz
Voltage 100-240VAC; 24V external transformer
Electrical Requirements 100-240VAC; 24V external transformer 50/60Hz
Includes Top and bottom probes, incubation to 50°C, shaking, 485 and 360nm excitation filters, 528 and 460nm emission filters, serial and power cables, power cord
Light Source Quartz halogen, 1000-hour life
Detector Type Fluorescence, Luminescence

  • All models read 6-, 12-, 24-, 48-, 96-, and 384-well plates
  • Onboard software stores 55 programmable protocols, up to 75 custom programs, and eight microplate assay results
  • Two-line, 24-character LCD and menu-driven software simplify operation and programming
  • Onboard diagnostic testing utilizes three methods — an optical self-test, a universal test plate, and a “wet test” — to check instrument performance and optical specifications, and provides either soft or hardcopy results
  • Fiber-optic probes provide excellent sensitivity over a wide range
  • All units feature convenient front-panel access to accessories

Software Features

  • Includes Gen5RC reader control software or Gen5 Data Analysis software (model dependent)
  • Onboard data analysis provides curve-fitting options, including linear regression, polynomial, point-to-point, cubic spline, and four-parameter logistic curve fits

Model Features

  • Top probe is ideal for reading solid black plates
  • Bottom probe reads covered plates
  • Combination of top and bottom probes offers the most flexibility and includes automated probe switching for added convenience
  • Extended-range photomultiplier tube (PMT) extends emission range to 800nm Shaking and incubation options are ideal for temperature-sensitive assays
  • Single reagent injector is suitable for ion-uptake assays and flash luminescence

Note
Supplied filters can be exchanged for other filters at time of purchase at no extra cost so that you can configure the unit to meet your specific application

Optional Gen5 software automatically runs the reader, analyzes the results, and generates a customized report. Or choose Gen5 Secure software to meet requirements of 21 CFR Part 11.

Certifications

CE marked and ETL listed, Certified to CAN/CSA C22.2 No. 1010.1-92 and EN61010-1, including amendments 1 and 2

Conforms to UL 3101-1