Please login to your online account to display your discounted pricing

BioTek™ Epoch™ 2 Microplate Spectrophotometer


Fully contained workstation offers a variety of data output options including Wi-Fi, Bluetooth and USB flash drive. BioTek Epoch 2 Microplate Spectrophotometer is a compact, monochromator-based microplate spectrophotometer combines modern touchscreen technology with excellent performance for UV-Vis measurements in 6- to 384-well microplates, cuvettes and in micro-volume samples with the available Take3™ plate.

Manufacturer: biotek™  EPOCH2

 View more versions of this product

Catalog No. BTEPOCH2

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Weight (Metric) 9.1kg
Electrical Requirements 100-240V 50/60Hz
Includes No Cuvette Port
Wavelength Repeatability ±2nm
Scan Speed 96 wells sweep read: 8 sec.; 384 wells sweep: 14 sec.
Wavelength Accuracy ±2nm
Voltage 100-240V
Hertz 50/60Hz
For Use With (Equipment) 6-, 12-, 24-, 48-, 96-, and 384-well microplates
Spectral Bandwidth 5nm
Lamp Xenon Flash Lamp

Comes with full-function Gen5™ software to read and analyze data. For maximum versatility, Epoch 2 can read from 200nm to 999nm in 1nm increments for single, dual and multi-wavelength measurements in end point and kinetic read methods.

 

  • UV-Vis wavelength selection in 2μL micro-volume samples, 6- to 384-well plates and standard cuvettes
  • Large color touchscreen with available Wi-Fi, Bluetooth and flash drive connectivity
  • Full Gen5™ data analysis onboard
  • Read modes include end point, kinetic, spectral scanning, and well area scanning
  • Advanced 4-Zone™ incubator design to 65°C with condensation control
  • Shaking profiles including linear, orbital and double orbital
  • Compatible with BioStack microplate stacker and third party automation
  • Other labware: Take3 and Take 3 Trio Microvolume Plates (optional); Bio-Cell™ (optional); Standard cuvette (with “C” configurations)
Specifications
  • Microplate types: 6-, 12-, 24-, 48-, 96-, and 384-well microplates
  • Wavelength selection: Monochromator
  • Wavelength range: 200 - 999nm, selectable in 1nm increments
  • Light source: Xenon flash lamp
  • Dynamic range: 0.0- 4.0 OD
  • Bandpass: 5nm
  • Shaking: Orbital, double orbital and linear
  • Temperature control: Ambient +4° to 65°C with condensation control
  • Connectivity: USB for external PC control; USB for peripheral devices
  • Power: 100-240V 50/60Hz
  • Dimensions (Touchscreen model): 39.3D x 32W x 33cmH (15.5 x 12.5 x 13 in.)
  • Dimensions (Non-Touchscreen): 39.3D x 32W x 20.3cmH (15.5 x 12.5 x 8 in.)
  • Weight (Touchscreen model): 25 lb. (11.3 kg); Non-touchscreen: 20 lb. (9.1kg)
Applications
  • Nucleic acid and protein direct quantification
  • Micro-volume assays with Take3™ plates
  • Biofuel research applications
  • Cytotoxicity assay
  • Cell proliferation assay
  • End point or kinetic ELISA
  • Spectral scanning

Certifications

CE and TUV marked