Please login to your online account to display your discounted pricing

BioTek™ Eon™ Microplate Spectrophotometers

High-performance spectrophotometers with filterless, continuous wavelength selection from 200 to 999nm

Specifications

Certifications/Compliance CE and TUV
Connections USB for PC connection
Depth (English) 16 in.
Depth (Metric) 40.6cm
Lamp Xenon
View More Specs
Includes:  Gen5™ v 2.0 software

 Disclaimers

For in vitro diagnostic use

Products
Catalog Number Mfr. No. Price Quantity    

11-120-611

 
biotek™
EON
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

11-120-612

 
biotek™
EONC
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Certifications/Compliance CE and TUV
Connections USB for PC connection
Depth (English) 16 in.
Depth (Metric) 40.6cm
Lamp Xenon
For Use With (Application) Nucleic acid quantification, protein quantification, micro-volume assays (with Take3™), biofuel research applications, cell proliferation, cytotoxicity, ELISA, enzyme kinetics
Height (English) 8.5 in.
Height (Metric) 21.6cm
Hertz 50/60Hz
Voltage 100 to 240V
Wavelength Accuracy ±2nm
Wavelength Range 200 to 999nm
Wavelength Repeatability 0.2nm
Weight (English) >24 lb.
Weight (Metric) >11kg
Width (English) 8.5 in.
Width (Metric) 21.6cm

Accommodate a wide range of assay applications in the life science laboratory, from DNA, RNA and protein quantification to long-running kinetic assays with monochromator-based optics, extended incubation range, optimized shaking capability, and optional cuvet port.

  • Read 6- to 384-well microplate formats
  • Optical system allows wavelength selection between 200 and 999nm in 1nm increments
  • 11-120-612 offers cuvet port (200 to 900nm)
  • View peak sample wavelengths using a quick spectral scan in a cuvet or microplate well
  • Multiple shake modes — choose orbital, double orbital or linear shaking with customizable shaking frequency and duration
  • 4-Zone™ temperature control system, plus 65°C setpoint to expand range of applications
  • Compatible with the Take3™ Micro-Volume plates for 2μL measurements
  • Compatible with BioStack™ automation

Software supports increased laboratory efficiency

  • Logical interface for easy flow: from reading parameters, plate layout, data reduction to flexible data output options
  • Intuitive design: StepWise™ protocol, powerful data reduction tools

Applications:

Nucleic acid quantification, protein quantification, micro-volume assays (with Take3™), biofuel research applications, cell proliferation, cytotoxicity, ELISA, enzyme kinetics

CE and TUV marked