Please login to your online account to display your discounted pricing

BioTek™ Eon™ Microplate Spectrophotometers


High-performance spectrophotometers with filterless, continuous wavelength selection from 200 to 999nm

Manufacturer: biotek™  EONC

 View more versions of this product

Catalog No. 11-120-612

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Certifications/Compliance CE and TUV
Connections USB for PC connection
Depth (English) 16 in.
Depth (Metric) 40.6cm
Detection Range 200 to 999nm, selectable in 1nm increments
Electrical Requirements 100 to 240V 50/60Hz
For Use With (Application) Nucleic acid quantification, protein quantification, micro-volume assays (with Take3™), biofuel research applications, cell proliferation, cytotoxicity, ELISA, enzyme kinetics
Height (English) 8.5 in.
Height (Metric) 21.6cm
Hertz 50/60Hz
Lamp Xenon
Voltage 100 to 240V
Wavelength Accuracy ±2nm
Wavelength Range 200 to 999nm
Wavelength Repeatability 0.2nm
Weight (English) >24 lb.
Weight (Metric) >11kg
Width (English) 8.5 in.
Width (Metric) 21.6cm

Accommodate a wide range of assay applications in the life science laboratory, from DNA, RNA and protein quantification to long-running kinetic assays with monochromator-based optics, extended incubation range, optimized shaking capability, and optional cuvet port.

  • Read 6- to 384-well microplate formats
  • Optical system allows wavelength selection between 200 and 999nm in 1nm increments
  • 11-120-612 offers cuvet port (200 to 900nm)
  • View peak sample wavelengths using a quick spectral scan in a cuvet or microplate well
  • Multiple shake modes — choose orbital, double orbital or linear shaking with customizable shaking frequency and duration
  • 4-Zone™ temperature control system, plus 65°C setpoint to expand range of applications
  • Compatible with the Take3™ Micro-Volume plates for 2μL measurements
  • Compatible with BioStack™ automation

Software supports increased laboratory efficiency

  • Logical interface for easy flow: from reading parameters, plate layout, data reduction to flexible data output options
  • Intuitive design: StepWise™ protocol, powerful data reduction tools

Applications:

Nucleic acid quantification, protein quantification, micro-volume assays (with Take3™), biofuel research applications, cell proliferation, cytotoxicity, ELISA, enzyme kinetics