missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BioTek™ ELx808™ Absorbance Microplate Readers

Store up to 55 user-programmable protocols. BioTek ELx808 Absorbance Microplate Readers read 96-well microplates and provide precise, accurate, and repeatable results.

Specifications

Accuracy >lt;1% at 2.5 O.D., >lt;2% at 3.5 O.D.
Measurement Speed 96 wells: 12 seconds (single wavelength), 20 seconds (dual wavelength), or 6 seconds (kinetic single wavelength)
Includes Filters, tungsten halogen light source, parallel printer cable, null modem serial cable and Gen5RC reader control software
Linearity >lt;1% at 2.5 O.D. (typical)
Weight (English) 30 lb.
View More Specs

Includes: Filters, tungsten halogen light source, parallel printer cable, null modem serial cable and Gen5RC reader control software


Products ${productFamilyLength}
Catalog Number Mfr. No. Wavelength Range Price Quantity  
Catalog Number Mfr. No. Wavelength Range Price Quantity  
BT-ELX808IU BioTek™
ELX808IU
340 to 900nm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT-ELX808 BioTek™
ELX808
380 to 900nm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Readers can perform multiple assays per plate
  • On-board diagnostic self-test may be used with an optional calibration test plate to easily confirm and document performance
  • Kinetic rate, R2, or onset time is reported without a PC
  • Linear well scanning is ideal for agglutination assays
  • Staggered optical design eliminates channel-to-channel crosstalk
  • Reference channel rolls optical signal for uniform channel-to-channel signal
  • Curve-fitting options include linear, 4-P, 2-P (logit/log), cubic, quadratic, cubic spline, and point-to-point
  • Measurements can be read in intervals as short as six seconds
  • Additional data analysis options include cutoff formulas, endpoint, and kinetic analysis Supplied with 4-Zone™ Temperature Control
  • Extended dynamic range

Model ELx808

  • Data analysis options include data transformation formulas, assay validation, and control validation
  • Optional 4-Zone™ Temperature Control improves reproducibility in enzyme kinetic reactions

Order Info

Optional Gen5 Data Analysis Software (Mfr. No. GEN5) provides simple, fast data collection, calculation, exporting, and reporting. Or select Gen5 Secure software to meet requirements of 21 CFR Part 11.

Certifications

CE marked

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

>lt;1% at 2.5 O.D., >lt;2% at 3.5 O.D.
Filters, tungsten halogen light source, parallel printer cable, null modem serial cable and Gen5RC reader control software
30 lb.
96 wells: 12 seconds (single wavelength), 20 seconds (dual wavelength), or 6 seconds (kinetic single wavelength)
>lt;1% at 2.5 O.D. (typical)
13.6kg