missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BioTek™ ELx800™ Absorbance Microplate Readers

Store up to 75 custom assay definitions and eight test results.

$9,693.87 - $9,693.87

Specifications

Accuracy ±1%, ±0.010Abs (from 0 to 2.000Abs @405nm)
Measurement Speed 96 wells: 30 seconds
Electrical Requirements 100-240V 50/60Hz
Hertz 50/60Hz
Linearity ±1% (from 0 to 2.000Abs @405nm), ±3% (from 2.000 to 3.000Abs @405nm)
View More Specs

Includes: Gen5RC reader control software


Products ${productFamilyLength}
Catalog Number Mfr. No. Accepts Model Wavelength Range Price Quantity  
Catalog Number Mfr. No. Accepts Model Wavelength Range Price Quantity  
BT-ELX800UV BioTek™
ELX800UV
340 to 750nm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT-ELX800NB BioTek™
ELX800NB
400 to 750nm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT-ELX800 BioTek™
ELX800
400 to 750nm Each for $9,693.87
Specifications

Description

  • Two 24-character LCD and convenient soft-keys for easy use
  • Onboard data analysis offers extensive curve-fitting options: linear, cubic, quadratic, 4-P, log-logit, cubic-spline, and point-to-point; plus endpoint and kinetic measurement
  • Three modes of performance and optical specifications testing: optical self-test, a calibration test plate, and a “wet test”
  • Auto or manual plate layout for blanks, controls, standards, and samples
  • Compatible with optional Gen5™ software
  • USB and RS232 ports for PC connection; parallel printer port
Features
  • ELx800 has 24-, 48-, and 96-well microplate read modes
  • ELx800UV contains optics to extend wavelength range down to 340nm
  • ELx800NB has the ability to read 384-well and 60-/72-/96-well Terasaki plates (with the help of optional software, sold separately)

Order Info

Optional Gen5 Data Analysis Software (Mfr. No. GEN5) provides simple, fast data collection, calculation, exporting, and reporting. Or choose Gen5 Secure software to meet requirements for 21 CFR Part 11 (Mfr. No. GEN5SECURE). 100-240VAC 50/60Hz.

Certifications

CE listed; ETL listed

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

±1%, ±0.010Abs (from 0 to 2.000Abs @405nm)
100-240V 50/60Hz
±1% (from 0 to 2.000Abs @405nm), ±3% (from 2.000 to 3.000Abs @405nm)
96 wells: 30 seconds
50/60Hz
100-240V