Please login to your online account to display your discounted pricing

BioTek™ ELx50™ Microplate Strip Washer


Syringe-drive fluid delivery system for precise control

Manufacturer: biotek™  ELX5012

 View more versions of this product

 Disclaimers

For in vitro diagnostic use

Catalog No. BT-ELX50-12

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Soak Time 1 to 600 sec.
Automation Interface RS-232
Width (Metric) 35cm
Weight (Metric) 8.9kg
Includes QC report and performance verification data sheet; Aerosol cover; Built-in pumps; Accessory package including bottles, tubing, and maintenance kit
Volume (Metric) Dispense 25 to 3000μL/well
Volume (Metric) Residual <4μL/well
Certifications/Compliance CE and ETL marked
Well Count 12
Wash Cycles 1 to 10
Display LCD
Width (English) 14 in.
Depth (Metric) 40cm
Depth (English) 16 in.
Height (Metric) 16.25cm
Height (English) 6.5 in.
Weight (English) 20 lb.
Hertz 50/60Hz
Type Standard Strip Washer
Voltage 100 to 250VAC
Flow Rate 150 to 1000μL/well/sec.
Electrical Requirements 100 to 250VAC 50/60Hz
For Use With (Application) Suitable for EIA, fluorescence, chemiluminescence, RIA, DNA probe, and cellular assays