Please login to your online account to display your discounted pricing

BioTek™ ELx50™ Microplate Strip Washer

Syringe-drive fluid delivery system for precise control

Specifications

Certifications/Compliance CE and ETL marked
Automation Interface RS-232
Depth (English) 16 in.
Depth (Metric) 40cm
Height (English) 6.5 in.
View More Specs
Includes:  QC report and performance verification data sheet; Aerosol cover; Built-in pumps; Accessory package including bottles, tubing, and maintenance kit

 Disclaimers

For in vitro diagnostic use

Products
Catalog Number Mfr. No. Type Well Count Price Quantity    

BT-ELX5016V

 
biotek™
ELX5016V
Washer with valves and automatic buffer switching 16 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BT-ELX50-12

 
biotek™
ELX5012
Standard Strip Washer 12 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BT-ELX50-8

 
biotek™
ELX508
Standard Strip Washer 8 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BT-ELX5016

 
biotek™
ELX5016
Standard Strip Washer 16 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BT-ELX50-12V

 
biotek™
ELX5012V
Washer with valves and automatic buffer switching 12 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BT-ELX50-8V

 
biotek™
ELX508V
Washer with valves and automatic buffer switching 8 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTELX508MF

 
biotek™
ELX50/8MF
Plate/Strip Washer w/vacuum filtration 96 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTELX508M

 
biotek™
ELX50/8M
Washer w/biomagnetic separation 96 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTELX508F

 
biotek™
ELX50/8F
Washer w/vacuum filtration 96 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Certifications/Compliance CE and ETL marked
Automation Interface RS-232
Depth (English) 16 in.
Depth (Metric) 40cm
Height (English) 6.5 in.
Height (Metric) 16.25cm
Includes QC report and performance verification data sheet; Aerosol cover; Built-in pumps; Accessory package including bottles, tubing, and maintenance kit
Soak Time 1 to 600 sec.
Wash Cycles 1 to 10
Weight (Metric) 8.9kg
Width (English) 14 in.
Display LCD
Electrical Requirements 100 to 250VAC 50/60Hz
Flow Rate 150 to 1000μL/well/sec.
For Use With (Application) Suitable for EIA, fluorescence, chemiluminescence, RIA, DNA probe, and cellular assays
Hertz 50/60Hz
Voltage 100 to 250VAC
Volume (Metric) Dispense 25 to 3000μL/well
Volume (Metric) Residual <4μL/well
Weight (English) 20 lb.
Width (Metric) 35cm

Key Features

  • Washes single strips or full plates
  • Efficient washing ensures low background absorbance and small residue volumes
  • Self-contained and programmable
  • Micro-controlled, syringe-drive positive-displacement pump fluid delivery system for precise control
  • Biomagnetic separation and vacuum filtration options available
  • Safety features include aerosol cover, built-in decontamination routine, and isolation of electronics from fluidics
  • Biomagnetic separation and vacuum filtration options available
  • Choice of 8-, 12- or 16-channel manifold models
  • Optional switching of up to three wash buffers

Applications:

Suitable for EIA, fluorescence, chemiluminescence, RIA, DNA probe, and cellular assays

CE listed; ETL listed

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.