Please login to your online account to display your discounted pricing

BioTek™ ELx50™ Microplate Strip Washer


Syringe-drive fluid delivery system for precise control

Manufacturer: biotek™  ELX50/8F

 View more versions of this product

 Disclaimers

For in vitro diagnostic use

Catalog No. BTELX508F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Certifications/Compliance CE and ETL marked
Weight (Metric) 8.9kg
Wash Cycles 1 to 10
Type Washer w/vacuum filtration
Height (English) 6.5 in.
Width (English) 14 in.
Includes QC report and performance verification data sheet; Aerosol cover; Built-in pumps; Accessory package including bottles, tubing, and maintenance kit
Volume (Metric) Dispense 25 to 3000μL/well
Weight (English) 20 lb.
Hertz 50/60Hz
Electrical Requirements 100 to 250VAC 50/60Hz
Flow Rate 150 to 1000μL/well/sec.
Well Count 96
Volume (Metric) Residual <4μL/well
Automation Interface RS-232
Soak Time 1 to 600 sec.
Depth (English) 16 in.
Display LCD
Width (Metric) 35cm
Height (Metric) 16.25cm
Voltage 100 to 250VAC
Depth (Metric) 40cm
For Use With (Application) Suitable for EIA, fluorescence, chemiluminescence, RIA, DNA probe, and cellular assays