BioTek™ ELx405™ Select Deep-Well Microplate Washer
World's first deep well microplate washer that aspirates and dispenses into 96- and 384-well plates <50mm in height
Manufacturer:  BioTek™ ELX405UVSD
 View more versions of this product
Catalog No. ELX405UVSD
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Certifications
CE and ETL marked
Specifications
Specifications
Built-in ultrasonic bath for protein and salt crystal buildup; programmable up to 60 minutes (model dependent)
Programmable in minutes and seconds, up to 60 min.
35.6cm
43.2cm
25.4cm
13.5kg
Dual-Action 96-tube manifold, automatic 4-buffer switching and built-in ultrasonic cleaning
100 to 240V
CE and ETL marked
<8A
10 to 80%, noncondensing
59° to 86°F
≤3% CV
Backlit LCD
14 in.
17 in.
10 in.
30 lb.
50/60Hz
≤2μL/well
In Vitro diagnostic use
One positive displacement pump
15° to 30°C
100 to 240V 50/60Hz
Documents
For in vitro diagnostic use