Please login to your online account to display your discounted pricing

BioTek™ ELx405™ Select Deep-Well Microplate Washer


World's first deep well microplate washer that aspirates and dispenses into 96- and 384-well plates <50mm in height

Manufacturer: biotek™  ELX405UVD

 View more versions of this product

 Disclaimers

For in vitro diagnostic use

Catalog No. ELX405UVD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Temperature (Metric) Operating 59° to 86°F
Weight (Metric) 13.5kg
Humidity 10 to 80%, noncondensing
Certifications/Compliance CE and ETL marked
Fluid Delivery Type One positive displacement pump
Wash Cycles Built-in ultrasonic bath for protein and salt crystal buildup; programmable up to 60 minutes (model dependent)
Volume (Metric) Residual ≤2μL/well
Includes Dual-Action 96-tube manifold and automatic 4-buffer switching
Height (English) 10 in.
Voltage 100 to 240V
Weight (English) 30 lb.
Hertz 50/60Hz
Electrical Requirements 100 to 240V 50/60Hz
Temperature (English) Operating 15° to 30°C
Amperage <8A
Width (English) 14 in.
Soak Time Programmable in minutes and seconds, up to 60 min.
Depth (English) 17 in.
Display Backlit LCD
For Use With (Application) In Vitro diagnostic use
Height (Metric) 25.4cm
Precision ≤3% CV
Width (Metric) 35.6cm
Depth (Metric) 43.2cm