Please login to your online account to display your discounted pricing
BioTek™ Cytation™ 5 Cell Imaging Multi-Mode Reader with Promo Starter Set
For 6- to 384- well microplates, slides, Petri dishes and flasks. BioTek Cytation 5 Cell Imaging Multi-Mode Reader includes fluorescence, brightfield, H&E and phase contrast microscopy and upgradable multi-mode microplate reading which provides powerful imaging and quantitative data collection in one instrument.
Products
Catalog Number Mfr. No. For Use With (Application) Model Price Quantity    
BTCYT5FAVPM
biotek™
CYT5FAVPM
Fluorescence, brightfield, H&E imaging and filter-based fluorescence, luminescence and Alpha detection CYT5FAV CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT5FVPM
biotek™
CYT5FVPM
Fluorescence, brightfield, H&E imaging and filter-based fluorescence and luminescence detection CYT5FV CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT5MFVPM
biotek™
CYT5MFVPM
Fluorescence, brightfield, H&E imaging, plus monochromator and filter-based multi-mode detection CYT5MFV CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT5MVPM
biotek™
CYT5MVPM
Fluorescence, brightfield, H&E imaging and monochromator fluorescence, UV-Vis absorbance and luminescence CYT5MV CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT5PVPM
biotek™
CYT5PVPM
Fluorescence, brightfield, H&E and phase contrast imaging CYT5PV CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT5VPM
biotek™
CYT5VPM
Fluorescence, brightfield, H&E imaging CYT5V CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCY5MFAVPM
biotek™
CYT5MFAVPM
Fluorescence, brightfield, H&E imaging, monochromator and filter multi-mode, laser-based Alpha CYT5MFAV CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Special Promo Offer: Each reader comes with a Free Imaging Starter Set.
  • Six position objective turret; 2.5X to 60X magnification available
  • 4X, 10X, 20X and 40X phase contrast objectives available
  • Linear, orbital and double-orbital shaking
  • Live cell imaging and assay support with CO2/O2 control, incubation to 65°C and shaking optimize cell-based and other assays
  • Upgradable to add patented Hybrid Technology™ in multi-mode detection combines high performance filters with variable bandwidth monochromators for versatility and performance
  • The fluorescence monochromators offer a variable bandwidth from 9nm to 50nm in 1nm increments for ultimate flexibility
  • Laser-based excitation for AlphaScreen™/AlphaLISA™assays
  • CO2/O2 control, incubation to 65°C and shaking optimize cell-based and other assays
  • Low volume (2μL) nucleic acid and protein quantification with the available Take3™Plates
  • Powerful yet easy to use Gen5 software for efficient plate reading, image capture and analysis
Features
  • BTCYT5V: Fluorescence, brightfield and H&E imaging; incubation to 65°C and linear, orbital and double-orbital shaking
  • BTCYT5PV: Phase contrast, fluorescence, brightfield and H&E imaging; incubation to 65C and linear, orbital and double-orbital shaking
  • BTCYT5FV: Fluorescence, brightfield and H&E imaging, filter/dichroic fluorescence, fluorescence polarization, time resolved fluorescence, luminescence and filtered luminescence; incubation to 65C and linear, orbital and double-orbital shaking
  • BTCYT5FAV: Fluorescence, brightfield and H&E imaging, filter/dichroic fluorescence, fluorescence polarization, time resolved fluorescence, luminescence, filtered luminescence and laser AlphScreen/AlphaLISA; incubation to 65C and linear, orbital and double-orbital shaking
  • BTCYT5MV: Fluorescence, brightfield and H&E imaging with monochromator fluorescence, monochromator UV-Vis absorbance, time resolved fluorescence (secondary mode) and luminescence; incubation to 65C and linear, orbital and double-orbital shaking
  • BTCYT5MPV: Fluorescence, brightfield, phase contrast and H&E imaging with monochromator fluorescence, monochromator UV-Vis absorbance, time resolved fluorescence (secondary mode), luminescence; incubation to 65C and linear, orbital and double-orbital shaking
  • BTCYT5MFV: Fluorescence, brightfield and H&E imaging with monochromator fluorescence, filter/dichroic fluorescence, monochromator UV-Vis absorbance, fluorescence polarization, time resolved fluorescence, luminescence, filtered luminescence incubation to 65C and linear, orbital and double-orbital shaking
  • BTCYT5MFAV: Fluorescence, brightfield and H&E imaging with monochromator fluorescence, filter/dichroic fluorescence, monochromator UV-Vis absorbance, fluorescence polarization, time resolved fluorescence, luminescence, filtered luminescence and laser AlphaScreen/AlphaLISA; incubation to 65C and linear, orbital and double-orbital shaking