Please login to your online account to display your discounted pricing

BioTek™ Cytation™ 3 Multi-Mode Reader


Combines automated digital microscopy and conventional microplate detection. BioTek Cytation 3 Multi-Mode Reader with its unique design provides rich phenotypic cellular information with well-based quantitative data.

Manufacturer: biotek™  CYT3MF

 View more versions of this product

Catalog No. BTCYT3MF

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Monochromator, Fluorescence and Absorbance; Filter Based
Depth (Metric) 50.8cm
Width (Metric) 41.91cm
Width (English) 16.5 in.
Height (English) 17.5 in.
Depth (English) 20 in.
Height (Metric) 44.5cm

  • Automated digital microscopy and/or multi-mode microplate detection in one instrument
  • Modular and upgradable: microscopy only, microplate reading only, or both
  • Flexible sample format: 6- to 384-well plates, microscope slides, cell culture flasks and Petri dishes
  • Affordable automation: automated XY stage, auto focus, auto exposure, automated image capture, LED intensity
  • Inverted fluorescence and brightfield microscopy
  • High Power LED cubes that provide superior excitation energy and long life
  • Optimized epifluorescence filter cubes for superior sensitivity for commonly used fluorophores: DAPI, CFP, GFP, YFP, RFP, Texas Red, CY5 and CY7
  • From 2.5x for full-well imaging to 60x for sub-micron resolution for intra-cellular details
  • Live cell assays with temperature control up to 45°C, CO2/O2 gas control accessory
  • Endpoint, time-lapse and montage assays
  • Automatic cell counting, sub-population analysis and image statistics
  • Dual reagent injectors
  • Patented Hybrid Technology™ microplate reading mode with high performance filter-based optics and high-flexibility quadruple monochromator optics
Note: Models BTCYT3MF. BTCYT3M, BTCYT3F can be upgraded to add fluorescence and brightfield imaging. Other models are available that include fluorescence and brightfield imaging; see Cat. Nos. BTCYT3V, BTCYT3FV, BTCYT3MV, BTCYT3MFV.