Please login to your online account to display your discounted pricing

BioTek™ 405™ Touch Microplate Washer


Configured with many options to meet myriad assay requirements

Manufacturer: biotek™  405TSHTV

 View more versions of this product

Catalog No. BT405TSHTV

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Automation Interface High res LED backlit touch screen for instrument control
Assays ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes
Well Count 384
Height (Metric) 25.4cm
USB Connections 2
Volume (Metric) Wash 25 to 3000μL/well
Type Microplate Washer w/4-Buffer switching, ultrasonic advantage
Width (English) 14 in.
Height (English) 10 in.
Wash Cycles 1 to 250
Washing Speed Solid bottom plates: 3 asp./disp/ cycles, 300 uL/well, 96 wells, 96 tube manifold, final aspirate: <30s; 3 asp./disp/ cycles, 100 uL/well, 384 wells, 96 tube manifold, final aspirate: <80s; 1 asp./disp/ cycles, 40 uL/well, 384 wells, 192 tube manifold
Electrical Requirements 100 to 240V 50/60Hz
Volume (Metric) Residual ≤2μL/well
Voltage 100 to 240V
Fluid Delivery Type Internal positive displacement pump
Display LED
Flow Rate 1 to 11
Depth (English) 17 in.
Weight (Metric) With Internal Buffer Switching: 16.5kg; Without Internal Buffer Switching: 13.5kg
Soak Time Programmable in minutes and seconds, up to 60 min.
For Use With (Application) ELISA, MSD assays, HCS immune cytochemistry, FLIPR Ca2+ flux, Cell-based assays, Magnetic and polystyrene bead processing (gene expression assays, cytokine assays), ELISPOT assays
Width (Metric) 35.6cm
Hertz 50/60Hz
Depth (Metric) 43.2cm
Weight (English) With Internal Buffer Switching: 36 lb.; Without Internal Buffer Switching: 30 lb.