Please login to your online account to display your discounted pricing

BioTek™ 405™ LS Microplate Washers with Ultrasonic Advantage™

Fifth generation washer with patent-pending built-in Ultrasonic Advantage for automated manifold cleaning

Specifications

Humidity 10 to 85%, non condensing
Temperature (Metric) Operating 15° to 30°C
Voltage 100 to 240V AC
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    

BT405LSRS

 
biotek™
405LSRS
Washer for 96-well microplate washing CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BT405LSRVS

 
biotek™
405LSRVS
Washer for 96-well microplate washing with 4 buffer switching CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Humidity 10 to 85%, non condensing
Temperature (Metric) Operating 15° to 30°C
Voltage 100 to 240V AC

  • For easy processing of cell based assays, ELISA and multiplex assays
  • Angled dispense tubes, extensive aspirate and dispense rates, plate position controls provide optimal conditions for washing even fragile cell layers
  • Programmable in minutes and seconds, up to 60 minutes (variable intensities)
  • Waste and vacuum sensing, fluid and flow detection
  • Aerosol cover and overflow protection
Hardware
  • Accommodates 96 well microplates
  • 2 x 24 character backlit LCD display and membrane keypad simple user interface
  • Automated internal four buffer switching
  • Automates basic 96 well dispensing and aspiration
  • Internal positive displacement pump eliminates use of troublesome pressure delivery systems and allows use of any size dispense reservoir
  • Built-in priming trough allows automatic manifold priming and rinsing during both operation and maintenance modes
  • Quick change manifold design
  • BioStack and third party automation compatible
  • All washers are biomagnetic separation ready (accessory magnets sold separately)
  • All washers are vacuum filtration ready (accessory filtration modules sold separately)
Software
  • Quick menu allows fluid paths to be easily primed, wash process started and manifold cleaned
  • Predefined protocols included for out-of-box washing
  • Fully customizable protocol parameters for individualized wash requirements (gentle cell washing to vigorous wash for high binding assays)
  • AutoPrime and auto maintenance routines
  • Additional software flexibility and 21CFR Part 11 compliance with LHC™ software for method development, downloading and washing plates
  • Run protocols created onboard or downloaded from LHC software

CE Marked