BioTek™ 405™ LS Microplate Washers For 96 Well Microplate Washing
Fifth generation washer automates basic 96-well dispensing and aspiration
Specifications
Electrical Requirements 100 to 240V AC 50/60 Hz ≤8A
Humidity 10 to 85%, non condensing
Temperature (Metric) Operating 15° to 30°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
BT405LSR BioTek™
405LSR
Washer for 96-well microplate N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSRV BioTek™
405LSRV
Washer for 96-well microplate with four buffer switching N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
  • For easy processing of cell based assays, ELISA and multiplex assays
  • Angled dispense tubes, extensive aspirate and dispense rates, plate position controls provide optimal conditions for washing even fragile cell layers
  • Programmable in minutes and seconds, up to 60 minutes (variable intensities)
  • Waste and vacuum sensing, fluid and flow detection
  • Aerosol cover and overflow protection
Hardware
  • Accommodates 96 well microplates
  • 2 x 24 character backlit LCD display and membrane keypad user interface
  • Automated internal four buffer switching
  • Internal positive displacement pump eliminates use of troublesome pressure delivery systems and allows use of any size dispense reservoir
  • Built-in priming trough allows automatic manifold priming and rinsing during both operation and maintenance modes
  • Quick change manifold design
  • BioStack and third party automation compatible
  • All washers biomagnetic separation ready (accessory magnets sold separately)
  • All washers vacuum filtration ready (accessory filtration modules sold separately)
Software
Quick menu allows fluid paths to be easily primed, wash process started and manifold cleaned
Predefined protocols included for out-of-box washing
Fully customizable protocol parameters for individualized wash requirements (gentle cell washing to vigorous wash for high binding assays)
AutoPrime and auto maintenance routines
  • Run protocols created onboard or downloaded from LHC Software
  • Certifications
    CE Marked
    Description & Specifications
    Specifications
    100 to 240V AC 50/60 Hz ≤8A
    10 to 85%, non condensing
    15° to 30°C