missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide C18 HPLC Column, 160 Å, 4.7 μm Particle Size
BIOshell™ 160 Å Peptide C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins.
$871.10 - $1502.18
Specifications
Particle Size | 4.7 μm |
---|---|
pH | 1 to 8 |
Pore Size | 160 Å |
Surface Area | 60 m2/g |
Stationary Phase | C18 (octadecyl) |
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity | ||||
111008369
|
MilliporeSigma™ Supelco™
67001U |
2.1 mm | 3 cm |
Each for $871.10
|
|
||||
111007926
|
MilliporeSigma™ Supelco™
67002U |
2.1 mm | 5 cm |
Each for $1,015.03
|
|
||||
111007669
|
MilliporeSigma™ Supelco™
67003U |
2.1 mm | 7.5 cm |
Each for $1,191.75
|
|
||||
111007512
|
MilliporeSigma™ Supelco™
67004U |
2.1 mm | 10 cm |
Each for $1,304.15
|
|
||||
111007057
|
MilliporeSigma™ Supelco™
67006U |
2.1 mm | 15 cm |
Each for $1,502.18
|
|
||||
111008370
|
MilliporeSigma™ Supelco™
67007U |
3 mm | 3 cm |
Each for $871.10
|
|
||||
111007927
|
MilliporeSigma™ Supelco™
67008U |
3 mm | 5 cm |
Each for $1,015.03
|
|
||||
111007513
|
MilliporeSigma™ Supelco™
67011U |
3 mm | 10 cm |
Each for $1,304.15
|
|
||||
111007058
|
MilliporeSigma™ Supelco™
67012U |
3 mm | 15 cm |
Each for $1,502.18
|
|
||||
111007928
|
MilliporeSigma™ Supelco™
67013U |
4.6 mm | 5 cm |
Each for $1,015.03
|
|
||||
111007514
|
MilliporeSigma™ Supelco™
67014U |
4.6 mm | 10 cm |
Each for $1,304.15
|
|
||||
111007059
|
MilliporeSigma™ Supelco™
67015U |
4.6 mm | 15 cm |
Each for $1,502.18
|
|
||||
Description
BIOshell™ 160 Å Peptide C18 columns contain advanced Fused-Core particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping. Additionally, the sterically-protected C18 ligands provide extra stability allowing the columns to be used at an extended pH range (2-9) and high temperatures (up to 90°C). This greatly expands the application range for the separation of biomoleculesSpecifications
4.7 μm | |
160 Å | |
C18 (octadecyl) | |
C18 (octadecyl) Phase/Fused-Core Particle Platform | |
No | |
Stainless Steel |
1 to 8 | |
60 m2/g | |
BIOshell | |
4% | |
Reversed Phase | |
Fused-Core HPLC |
BIOshell is a trademark of Sigma-Aldrich Co. LLC