Bioptechs™ Delta T Culture Dishes
For used in fluorescence, DIC, and confocal modes of microscopy, or anywhere that highest image quality is desirable
Manufacturer: Bioptechs™ 04200417C
Description
- Disposable plastic culture dish with the high optical compatibility of glass
- Contain self heating ITO coating, which provides first surface heat transfer directly to cells when using Delta T System
- Injection molded polystyrene sidewalls and lid, coverglass bottom
- Highest quality German Desag 263, low alkaline coverglass for optimum resolution
- 35mm O.D. and 23mm central aperture with tapered sidewalls and low profile (6mm in height) to reduce dead-volume and allow better access for microinjection and micromanipulation
- Glass thicknesses of 0.17mm
- Available in clear and black (for fluorescence applications)
- Numerous accessories specific to these dishes allow for specialized applications including - media perfusion, gas perfusion, sample positioning, heating and cooling
- Optional stage adapters available to fit all major brands of microscopes, which custom stage adapters available
- Optional culturing cylinders may be used to barricade cells or suspended specimens in a dish; or to restrict and concentrate growth and location of cells plated on coverslip without use of grease
Specifications
Clear |