Thermo Scientific™ BioLite Filter Units
Safeguard your cells and your budget. Choose economical filter units with aPES membranes for performing general filtration procedures for sterile filtration of your cell culture media.
$112.07 - $328.34
Specifications
Pore Size 0.2µm
Sterile Yes
Membrane PES
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size Volume (Metric) Receiver Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size Volume (Metric) Receiver Price Quantity  
12-566-215 Thermo Fisher Scientific
N130195
50mm 0.2µm 150mL Case of 12 Each for $112.07
12-566-218 Thermo Fisher Scientific
N130199
90mm 0.2µm 1000mL Case of 12 Each for $328.34
12-566-217 Thermo Fisher Scientific
N130197
75mm 0.2µm 500mL Case of 12 Each for $242.82
12-566-216 Thermo Fisher Scientific
N130196
50mm 0.2µm 250mL Case of 12 Each for $144.52
Specifications
Description
Thermo Scientific™ BioLite Filter Units are a cost-effective filtration solution for cell culture applications where low protein binding is required. BioLite Filter Units are available in four volume sizes -150mL, 250mL, 500mL, and 1000mL - with 0.2 micron aPES membranes for removal of bacteria. All filter units are sterilized and individually bagged; each unit includes a sterilized closure cap that securely seals filtered media in receiver bottle for short term storage.
  • Safeguard your samples from contamination with 0.2 micron filter membranes for removal of bacteria
  • aPES membranes suited for low protein binding to maintain protein balance, and low extractables to retain media purity
  • BioLite filter top unscrews from storage bottle; a storage bottle cap is included with each filter unit for storing media
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation-sterilized and individually bagged
  • Note: Certificates of Processing and Certificates of Conformance are not available.
Description & Specifications
Specifications
0.2µm
Yes
PES