Please login to your online account to display your discounted pricing

Sartorius™ Optifit Tips in FlexiBulk Packs

Combine cost savings with saving space, improving usability and ensuring purity. Sartorius™ Optifit Tips in FlexiBulk Packs deliver the same high-quality Optifit Tips for use with Biohit™ pipettes in a convenient packaging design. Available in 200, 350, 1000 and 1200μL tip volumes, including a Wide Bore option.

$60.53 - $60.53

Specifications

For Use With (Equipment) Sartorius™ Biohit™ and many other brands of pipettes
Certifications/Compliance Certified to be DNase, RNase and Endotoxin Free
ISO 9001, ISO 14001 and ISO 13485 compliant
For Use With (Application) For everyday laboratory work
Packaging FlexiBulk
View More Specs
Products
Catalog Number Mfr. No. Color Code Length (Metric) Volume (Metric) Price Quantity    

14-559-490

 
sartorius™
791204
Purple 7.15cm 50 to 1200μL Pack of 1000 Each for $60.53
Case of 8 Pack for $484.24
Description & Specifications

Specifications

For Use With (Equipment) Sartorius™ Biohit™ and many other brands of pipettes
Certifications/Compliance Certified to be DNase, RNase and Endotoxin Free
ISO 9001, ISO 14001 and ISO 13485 compliant
For Use With (Application) For everyday laboratory work
Packaging FlexiBulk

Optifit Tips

  • Perfect fit and seal to ensure the highest possible accuracy and precision
  • Ergonomic and light tip attachment/ejection via the Optiload feature in Sartorius pipettes
  • Color coded for easy matching with suitable Sartorius pipettes
  • Wide bore tips available for better dispensing of difficult samples, such as viscous liquids
  • Compatible with Picus™ NxT, Picus™, eLINE™, mLINE™ and Proline™ Plus pipettes and most other pipette brands
  • Compliant with ISO 9001, ISO 14001 and ISO 13485 quality standards
FlexiBulk Packaging
  • Convenience and savings
    • Combines the low cost of bulk tips with saving space and improving usability
    • Reduces size of packaging up to 40% and cuts packaging waste by nearly 50%
    • Materials in the FlexiBulk pack are 100% suitable to be used as energy waste
    • Pick & Place organization for quick and easy rack loading direct tip attachment
    • Easy open/close lid for easy tip access and for protecting tips
  • Quality and purity
    • Includes the exact same, high-quality tips as other packaging options
    • Ensures tip purity with lot-specific certificates
    • Nonsterile tips are certified free of RNase, DNase and endotoxins
    • Manufactured under ISO Class 8 cleanroom conditions
    • Lid provides an airtight seal during transport and storage for secured purity