Please login to your online account to display your discounted pricing

Biodiesel Fuel Standard, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$300.17 - $1,543.66

Chemical Identifiers

Chemical Name or Material Biodiesel Fuel Standard in Pyridine
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
View More Specs
Includes:  Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BD3508

 
spex certiprep llc
SPXBD3508
2mL amber prescored glass ampule 1mL Each for $1,543.66

BD3509

 
spex certiprep llc
SPXBD3509
2mL amber prescored glass ampule 1mL Each for $300.17
Description & Specifications

Specifications

Quantity 1mL
Chemical Name or Material Biodiesel Fuel Standard in Pyridine
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Packaging 2mL amber prescored glass ampule
Recommended Storage Freezer
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
DOT Information Nonhazardous
Solvent or Matrix Pyridine
Physical Form Fluid
CAS Min % 0.8
CAS Max % 99.2
CAS 122-32-7
CAS 56-81-5
CAS 621-71-6
CAS Max % 0.8
CAS 1718-52-1
CAS Min % 99.2

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Freezer