BioCision™ TruCool™ Cryogenic Vial Auto-cap Inserts
For use with TruCool™ Leak-proof cryogenic vials
$102.44 - $102.44
Specifications
Type Cryogenic Vial Auto-cap Insert
For Use With TruCool™ Auto-cap cryogenic vials
For Use With (Application) Vapor phase liquid nitrogen (LN2) only
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
13-900-885 BioCision™
MCS2435
White Pack of 1000 Each for $102.44
13-900-880 BioCision™
MCS2430
Blue Pack of 1000 Each for $102.44
13-900-882 BioCision™
MCS2432
Pink Pack of 1000 Each for $102.44
13-900-881 BioCision™
MCS2431
Green Pack of 1000 Each for $102.44
13-900-883 BioCision™
MCS2433
Red Pack of 1000 Each for $102.44
13-900-884 BioCision™
MCS2434
Yellow Pack of 1000 Each for $102.44
13-900-886 BioCision™
MCS2436
Violet Pack of 1000 Each for $102.44
13-900-887 BioCision™
MCS2437
Orange Pack of 1000 Each for $102.44
13-900-888 BioCision™
MCS2438
Gray Pack of 1000 Each for $102.44
Specifications
Description
  • Screw cap with thermally-fused gasket to ensure tight seal
  • Gasket is co-molded thermoplastic elastomer (TPE) layer that prevents leaking, slipping and risk of contamination
  • 95kPa certified gasket provides leak-proof seal
  • Star shaped caps available in different colors
  • Snap into open cavity of internal and external-threaded screwcap
Description & Specifications
Specifications
Cryogenic Vial Auto-cap Insert
TruCool™ Auto-cap cryogenic vials
Vapor phase liquid nitrogen (LN2) only