BioCision™ TruCool™ Auto-Cap Cryogenic Vials - Internal Threads
Recommended for use in vapor phase liquid nitrogen only
$480.38 - $783.89
Specifications
Diameter (Metric) Outer 12.5mm
Sterility Sterile
Thread Style Internal
For Use With (Application) Cell culture and biobanking
Includes Screw cap with thermally-fused gasket
View More Specs
Products 7
Catalog Number Mfr. No. Height (English) Height (Metric) Volume (Metric) Working Price Quantity  
Catalog Number Mfr. No. Height (English) Height (Metric) Volume (Metric) Working Price Quantity  
13-901-242 View Documents BioCision™
MCS2511
1.8″ 48mm 2mL Case of 500 Each for $513.90
13-901-241 View Documents BioCision™
MCS2510
1.69″ 43mm 1mL Case of 500 Each for $480.38
13-901-243 View Documents BioCision™
MCS2512
1.85″ 47mm 2mL Case of 500 Each for $480.38
13-901-244 View Documents BioCision™
MCS2513
2.9″ 75mm 4mL Case of 500 Each for $672.16
13-901-245 View Documents BioCision™
MCS2514
3.7″ 76.5mm 4mL Case of 500 Each for $672.16
13-901-246 View Documents BioCision™
MCS2515
3.52″ 89.5mm 5mL Case of 500 Each for $783.89
13-901-247 View Documents BioCision™
MCS2516
3.56″ 90.5mm 5mL Case of 500 Each for $783.89
Description
Description
  • Leak-proof cryogenic vial features novel screw cap with thermally-fused gasket to ensure tight seal
  • Made of medical grade polypropylene, will not discolor when re-sterilized
  • Each vial is individually barcoded with unique identifier that can be read with common barcode readers
  • Sterile vial with large white writing area
  • Graduations in 0.5mL increments
  • Non-cytotoxic, non-pyrogenic vial
  • Tube bottom works with interlocking cryoracks
  • Gasket is co-molded thermoplastic elastomer (TPE) layer that prevents leaking, slipping and risk of contamination
  • 95kPa certified gasket provides leak-proof seal
Specifications
Specifications
12.5mm
Internal
Screw cap with thermally-fused gasket
500/Cs.
Sterile
Cell culture and biobanking
Auto-Cap Cryogenic Vial