Please login to your online account to display your discounted pricing
BioCision™ HistoMailer™
Embedded cassette shipper
$33.33 - $33.33
Specifications
For Use With For securing histology biopsy samples
Material Eco-friendly, Recycled Polyethylene Terephthalate (PET) Material
Size Hold four samples
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
13-900-716
biocision™
BCS300
10/Pk. Each for $33.33
Description & Specifications
Specifications
For Use With For securing histology biopsy samples
Material Eco-friendly, Recycled Polyethylene Terephthalate (PET) Material
Size Hold four samples
Developed by the Mayo Clinic, HistoMailers ensure safe shipping of four histology embedded biopsy cassettes.
  • Prevents opening of cassettes and cross-contamination during shipping
  • Minimizes misidentification or mix-ups; allows for individual labeling and clear identification of samples at all times
  • Ships at room temperature or colder (with ice or dry ice)
  • Made of eco-friendly, recycled PET material
  • Useful for long- or short-term storage